missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCP042012F
Change view
Click to view available options
Package: Fiber Drum; Size: 12 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RDCP042012F Package: Fiber Drum; Size: 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCP042012F Supplier Ricca Chemical Company Supplier No. RDCP042012F4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7778-50-9
CAS Min % 98.0
CAS Max % 100.0
Quantity 12 kg
Molecular Formula Cr2K2O7
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 294.182
PubChem CID 24502
ChEBI CHEBI:53444
Grade ACS
Packaging Fiber Drum
Chemical Name or Material Potassium Dichromate
Show More Show Less
ShelfLife 60 months