missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dichromate, 0.0250 N (N/40), Ricca Chemical
Specifications
Specifications
| CAS | 7732-18-5 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Methods | ASTM/EPA for Chemical Oxygen Demand (COD) |
| pH | ∼4 |
| Quantity | 500 mL |
| Chemical Name or Material | Potassium Dichromate |