missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, 0.0250 N (N/40), Ricca Chemical

Catalog No. 605016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
605016 500 mL Natural Poly Bottle
605032 1 L Natural Poly Bottle
60501 4 L Natural Poly Bottle
R605000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 605016 Supplier Ricca Chemical Company Supplier No. 605016
Only null left

For ASTM/EPA Testing

Specifications

CAS 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Methods ASTM/EPA for Chemical Oxygen Demand (COD)
pH ∼4
Quantity 500 mL
Chemical Name or Material Potassium Dichromate