missing translation for 'onlineSavingsMsg'
Learn More

Potassium D-gluconate, 99%

Catalog No. p-7028023
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2513518 50 g
AAB2513530 250 g
AAB251350B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2513518 Supplier Thermo Scientific Chemicals Supplier No. B2513518

CAS: 299-27-4 | C6H11KO7 | 234.245 g/mol

Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

Solubility
Soluble in water. (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 299-27-4
Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.245
MDL Number MFCD00064211
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
Synonym potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
PubChem CID 16760467
IUPAC Name potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]

Specifications

Melting Point ∼180°C (decomposition)
Quantity 50 g
Solubility Information Soluble in water. (50mg/ml).
Formula Weight 234.25
Percent Purity 99%
Chemical Name or Material Potassium D-gluconate
EINECSNumber 206-074-2
RTECSNumber LZ5230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only