Learn More
Medisca Potassium Citrate, USP (Dietary Supplement Grade)
Description
Potassium Citrate, USP comes as either transparent crystals or as a white, granular powder. It is deliquescent when exposed to moist air.
Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.
Alternative name(s): Potassium citrate monohydrate, tripotassium citrate
Specifications
Specifications
| Chemical Name or Material | Potassium Citrate, USP (Dietary Supplement Grade) |
| Packaging | Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 6100-05-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 324.41 |
| Quantity | 100 g |
| Grade | USP |