missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monohydrate, Crystal, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P126512KG

 View more versions of this product

Catalog No. 18612577


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

6100-05-6
7.5 to 9.0
99%
Poly Pail
1
12 kg
Reagent

Chemical Identifiers

6100-05-6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.