missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monohydrate, Crystal, 99%, Spectrum™ Chemical

Catalog No. p-7166053
Change view
Click to view available options
Quantity:
12 kg
125 g
2.5 kg
45 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18612574 125 g Poly Bottle
18612575 500 g Poly Bottle
18612576 2.5 kg Poly Pail
18612577 12 kg Poly Pail
18612578 45 kg Fiber Drum
5 options

Catalog No. 18612574

Supplier: Spectrum Chemical Mfg Cor P1265125GM

Only null left

Chemical Identifiers

CAS 6100-05-6

Specifications

pH 7.5 to 9.0
CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.