missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate solution, Volumetric, 1/30 M K2CrO4 (0.1N), Honeywell Fluka™

Volumetric, 1/30 M K2CrO4 (0.1N)

$397.05 - $397.05

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002601
View Documents
Honeywell Chemicals
351571L
1 L N/A
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

7789-00-6
194.19
XMXNVYPJWBTAHN-UHFFFAOYSA-N
24597
[K+].[K+].[O-][Cr]([O-])(=O)=O
CrK2O4
MFCD00011368
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
CHEBI:75249

Specifications

7789-00-6
CrK2O4
MFCD00011368
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
[K+].[K+].[O-][Cr]([O-])(=O)=O
194.19
CHEBI:75249
Glass bottle
1 L
K2CrO4
UN3082
XMXNVYPJWBTAHN-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)chromium
24597
194.19g/mol
Potassium chromate solution
Videos
Safety and Handling

Safety and Handling

P201-P273-P280-P308 + P313-P333 + P313-P391

SDS
missing translation for 'documents'

missing translation for 'documents'