missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate solution, Volumetric, 1/30 M K2CrO4 (0.1N), Solstice

Catalog No. 6002601
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002601 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002601 Supplier Solstice Supplier No. 351571L
Only null left

Volumetric, 1/30 M K2CrO4 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 1 L
Linear Formula K2CrO4
UN Number UN3082
IUPAC Name dipotassium;dioxido(dioxo)chromium
Formula Weight 194.19g/mol
Packaging Glass bottle
Chemical Name or Material Potassium chromate solution
Hazard Statement P201-P273-P280-P308 + P313-P333 + P313-P391