missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Solution, Conductance Standard C Acc. to ISO 7888, 0.001 M KCl, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 7447-40-7 |
| Concentration | 0.001 M KCl |
| Linear Formula | KCl |
| MDL Number | MFCD00011360 |
| Quantity | 250 mL |
| Formula Weight | 74.55 |