Potassium Chloride Reference Solution A, 111,342 μS/cm at 25°C, 1 M, Ricca Chemical
Supplier: Ricca Chemical Company 589332
Specifications
Potassium Chloride Reference Solution A | |
1 L | |
ASTM D 1125 for Conductivity |
Natural Poly Bottle | |
1M |