missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, puriss. p.a., Honeywell Fluka™
Puriss. p.a.,≥99.5% (AT)
$142.90 - $381.03
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| SMILES | [Cl-].[K+] |
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 |
Specifications
| 7447-40-7 | |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| ≥99.5% (AT) | |
| 1.98g/mL at 25°C |
| 250 g | |
| KCl | |
| NONH for all modes of transport | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55g/mol | |
| puriss. p.a. | |
| Potassium chloride |
Safety and Handling
RTECSNumber : TS8050000