missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E508, 99-100.5% (AT), ≤0.0001% Al
$148.35 - $1,569.63
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| SMILES | [Cl-].[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 |
Specifications
| 7447-40-7 | |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| 99 to 100.5% (AT) | |
| Plastic Bottle | |
| Potassium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC |
| 500 g | |
| KCl | |
| NONH for all modes of transport | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55g/mol | |
| puriss. | |
| 1.98g/mL at 25°C |
Safety and Handling
EINECSNumber : 231-211-8
RTECSNumber : TS8050000