missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015421
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015421 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015421 Supplier Merck Emd Millipore Supplier No. PHR13295G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 7447-40-7
Density 1.98 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula KCl
MDL Number MFCD00011360
Melting Point 770°C
Synonym Potassium chloride
Type Certified Reference Material
Quantity 5 g
Formula Weight 74.55
Grade Certified Reference Material
Show More Show Less