missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Potassium Chloride, Molecular Biology Grade, Calbiochem™,

Catalog No. p-5547644
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
529552250GM 250 g
5295521KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 529552250GM Supplier MilliporeSigma™ Supplier No. 529552250GM
Only null left

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Quantity 250 g
Assay Percent Range ≥99%
Solubility Information Easily soluble in cold water
Formula Weight 74.6
Chemical Name or Material Potassium Chloride
Grade by argentometry
RTECSNumber TS8050000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area.