missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Granular, BiotechGrade, 99-100.5%, Spectrum™ Chemical
P1251, 7447-40-7, KCl
$511.70 - $5207.47
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605507
|
Spectrum Chemical Mfg Cor
P1251500GM |
500 g | Poly Bottle |
Each for $511.70
|
|
||||
|
18605508
|
Spectrum Chemical Mfg Cor
P125125KG |
2.5 kg | Poly Bottle |
Each for $1,324.07
|
|
||||
|
18605509
|
Spectrum Chemical Mfg Cor
P125112KG |
12 kg | Poly Pail |
Each for $3,322.57
|
|
||||
|
18605510
|
Spectrum Chemical Mfg Cor
P125150KGBL |
50 kg | Fiber Drum |
Each for $5,207.47
|
|
||||