missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Grade, 99.5% min, MilliporeSigma™

Catalog No. M1049361000
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1049361000 1 kg
M1049365000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1049361000 Supplier Technidata Supplier No. 1.04936.1000
Only null left

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color White
Physical Form Crystalline Solid
Quantity 1 kg
Assay Percent Range 99.5% min.
Linear Formula KCl
Formula Weight 74.55
Density 1.98g/cm3 (20°C); Bulk Density: 1000 kg/m3
Chemical Name or Material Potassium Chloride, EMSURE™, For Analysis
Recommended Storage Store without restrictions