missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, extra pure

CAS: 7447-40-7 | ClK | 74.55 g/mol

$181.89 - $571.02

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC193780010
SDS View Documents
Thermo Scientific Chemicals
193780010
1 kg Plastic bottle
Each for $181.89
Only null left
 
AC193780050
SDS View Documents
Thermo Scientific Chemicals
193780050
5 kg Plastic bottle
Each for $571.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium;chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]

Specifications

1420.0°C
7447-40-7
5.0 to 8.5 (5%, H2O, 25°C)
1 kg
KCl
06,480
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.54
Extra Pure
Potassium chloride
770.0°C
White
Crystalline Powder and/or Crystals
ClK
MFCD00011360
15, 7742
Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
[Cl-].[K+]
74.55
CHEBI:32588
≥99.0%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-211-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only