missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride (Crystalline/USP/FCC), Fisher Chemical™

Catalog No. P330500
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P330500 500 g Poly Bottle
P3303 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P330500 Supplier Thermo Fisher Scientific Supplier No. P330 500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420°C
Melting Point 770°C
Color White
pH 6
Physical Form Solid
Quantity 500 g
Formula Weight 74.55
Percent Purity 99.0 to 100.5% (USP)
Grade FCC/USP
Identification Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Loss on Drying 1.0% max. (FCC), 1.0% max. (USP), 1.0% max. (USP/FCC)
Packaging Poly Bottle
Chemical Name or Material Potassium Chloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:1