missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Specifications
Specifications
| Boiling Point | 1420°C |
| Melting Point | 770°C |
| Color | White |
| pH | 6 |
| Physical Form | Solid |
| Quantity | 10 kg |
| Assay Percent Range | 99.0 to 100.5 % |
| Formula Weight | 74.55 |
| Percent Purity | 99.0 to 100.5% |
| Grade | Certified ACS |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:1 Flammability:0 Instability:1 |