missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Crystal, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18605506
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
113.5 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605506 113.5 kg Fiber Drum
18605500 500 g Poly Bottle
18605502 2.5 kg Poly Bottle
18605501 6 x 500 g Poly Bottle
18605503 4 x 2.5 kg Poly Bottle
18605504 12 kg Poly Pail
18605505 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18605506 Supplier Spectrum Chemical Mfg Cor Supplier No. PO165113.5KGBL
Only null left

PO165, 7447-40-7, KCl

Spectrum™ Chemical Potassium Chloride, Crystal, USP is used to treat hypokalemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

CAS Max % 1
Quantity 113.5 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.01
Packaging Fiber Drum