missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 3000 μS/cm at 25°C, Ricca Chemical

Catalog No. R5888040500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5888040500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5888040500 Supplier Ricca Chemical Company Supplier No. R5888040500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 7447-40-7, 7732-18-5
CAS Min % 99.84, 0.16
CAS Max % 99.84, 0.16
Density 1.0011
Physical Form Liquid
Molecular Formula ClK
MDL Number MFCD00011360
Color Colorless
pH ∼7
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
Quantity 500 mL
PubChem CID 4873
ChEBI CHEBI:32588
Chemical Name or Material Potassium Chloride Conductivity Standard, 3000μS/cm at 25°C
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient