missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium chloride, for biochemistry

Catalog No. AC418205000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC418205000 500 g
AC418200025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418205000 Supplier Thermo Scientific Chemicals Supplier No. 418205000
Only null left

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for biochemistry

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Quantity 500 g
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information (1 M in water) Clear colorless
Formula Weight 74.54
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Potassium chloride
Show More Show Less
EINECSNumber 231-211-8

RUO – Research Use Only