missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, ACS Grade, 99.0 to 100.5%, LabChem™
Supplier: LabChem LC187901
Specifications
| 1,420°C | |
| 770°C | |
| White | |
| Crystals | |
| ClK | |
| MFCD00011360 | |
| Soluble in water | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| 99.0 to 100.5% | |
| Passes Test | |
| 1.987g/cm3 | |
| 1.987g/cm3 |
| Potassium Chloride | |
| 7447-40-7 | |
| 100 | |
| 500 g | |
| KCl | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55 | |
| ACS | |
| Poly Bottle | |
| Hydrogen chloride; Potassium oxide |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber : 231-211-8
Recommended Storage : Room Temperature