missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, ≥99%, MP Biomedicals

Catalog No. ICN19484494
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19484494 5 kg
ICN19484490 500 g
ICN19484491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19484494 Supplier MP Biomedicals Inc Supplier No. 0219484494
Only null left

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Quantity 5 kg
Assay Percent Range ≥99%
Formula Weight 74.55
Chemical Name or Material Potassium Chloride
Recommended Storage Room Temperature