missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium chloride, 99%, Molecular Biology Grade
CAS: 7447-40-7 | ClK | 74.55 g/mol
$285.99 - $451.92
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6418936
|
Thermo Scientific Chemicals
J6418936 |
500 g |
Each for $285.99
|
|
|||||
|
AAJ64189A1
|
Thermo Scientific Chemicals
J64189A1 |
1 kg |
Each for $451.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 |
Specifications
| 1500°C (sublimation) | |
| 7447-40-7 | |
| Powder | |
| ClK | |
| 14,7621 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium;chloride | |
| 4873 | |
| 74.55 | |
| Molecular Biology | |
| Hygroscopic | |
| Potassium chloride |
| 773°C | |
| White | |
| 500 g | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| 99% | |
| Odorless | |
| 1.984 g/mL |
Safety and Handling
EINECSNumber : 231-211-8
RTECSNumber : TS8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only