missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 99.999%, (trace metal basis)

Catalog No. AC268961000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268961000 100 g Plastic bottle
AC268960250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268961000 Supplier Thermo Scientific Chemicals Supplier No. 268961000
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99.999%
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
IUPAC Name potassium;chloride
Formula Weight 74.54
Percent Purity 99.999%
Grade Trace Metal Basis
Chemical Name or Material Potassium chloride
Show More Show Less
EINECSNumber 231-211-8

RUO – Research Use Only