missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 99.995% (metals basis)

CAS: 7447-40-7 | ClK | 74.55 g/mol

$247.14 - $937.38

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8762618
View Documents
Thermo Scientific Chemicals
08762618
50 g
Each for $247.14
Only null left
 
AA8762630
View Documents
Thermo Scientific Chemicals
08762630
250 g
Each for $937.38
Only null left
 
Description

Description

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
[Cl-].[K+]
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588

Specifications

1500°C (sublimation)
7447-40-7
Powder
99.995% (metals basis)
MFCD00011360
Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether.
[Cl-].[K+]
74.55
CHEBI:32588
Odorless
Hygroscopic
Potassium chloride
773°C
White
50 g
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium;chloride
4873
74.55
1.49
1.984 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 231-211-8

RTECSNumber : TS8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only