missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 2.00 M, Ricca Chemical

$331.97 - $554.84

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5899216
SDS View Documents
Ricca Chemical Company
5899216
500 mL Natural Poly Bottle
Each for $331.97
Only null left
 
589921
SDS View Documents
Ricca Chemical Company
589921
4 L Natural Poly Bottle
Each for $554.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

Colorless
Liquid
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.55
CHEBI:32588
Natural Poly Bottle
7732-18-5,7447-40-7
500 mL
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
Laboratory
Potassium Chloride
Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
missing translation for 'documents'

missing translation for 'documents'