missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol

Supplier:  Thermo Scientific Chemicals A170750I

 View more versions of this product

Catalog No. AAA170750I

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

Solubility
Soluble in water, glycerol. Insoluble in alcohol.

Notes
Store away from incompatible materials. Store in a well-ventilated place.
Specifications

Specifications

400°C (decomposition)
368°C
5000 g
MFCD00011361
14,7620
Soluble in water,glycerol. Insoluble in alcohol.
[O-]Cl(=O)=O.[K+]
122.55
122.55
Odorless
2.32
Potassium chlorate
3811-04-9
ClKO3
UN1485
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
VKJKEPKFPUWCAS-UHFFFAOYSA-M
potassium;chlorate
6426889
≥99%
Hygroscopic

Chemical Identifiers

3811-04-9
122.55
VKJKEPKFPUWCAS-UHFFFAOYSA-M
6426889
[O-]Cl(=O)=O.[K+]
ClKO3
MFCD00011361
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
potassium;chlorate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.

P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c

H271-H302+H332

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM CHLORATE

EINECSNumber : 223-289-7

RTECSNumber : FO0350000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only