missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%

Catalog No. p-7023481
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707530 250 g
AAA170750B 1000 g
AAA170750I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1707530 Supplier Thermo Scientific Chemicals Supplier No. A1707530

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol

Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

Solubility
Soluble in water, glycerol. Insoluble in alcohol.

Notes
Store away from incompatible materials. Store in a well-ventilated place.

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Boiling Point 400°C (decomposition)
Melting Point 368°C
Quantity 250 g
UN Number UN1485
Merck Index 14,7620
Solubility Information Soluble in water,glycerol. Insoluble in alcohol.
Formula Weight 122.55
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32
Chemical Name or Material Potassium chlorate
Show More Show Less
Hazard Category H271-H302+H332
Hazard Statement GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM CHLORATE
EINECSNumber 223-289-7
RTECSNumber FO0350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only