Learn More
Potassium carbonate sesquihydrate, ACS, 98.5-101.0%
For Manufacturing of Soaps, Pigments, Printing Ink | CAS: 6381-79-9 | C2H6K4O9 | 330.45 g/mol
$63.03 - $116.86
Chemical Identifiers
| CAS | 6381-79-9 |
|---|---|
| Molecular Formula | C2H6K4O9 |
| Molecular Weight (g/mol) | 330.45 |
| MDL Number | MFCD00149925 |
| InChI Key | WWVGGBSGYNQKCY-UHFFFAOYSA-J |
| IUPAC Name | tetrapotassium trihydrate dicarbonate |
| SMILES | O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O |
Description
Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
Chemical Identifiers
| 6381-79-9 | |
| 330.45 | |
| WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
| O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O |
| C2H6K4O9 | |
| MFCD00149925 | |
| tetrapotassium trihydrate dicarbonate |
Specifications
| 6381-79-9 | |
| 100 g | |
| C2H6K4O9 | |
| 14,7670 | |
| WWVGGBSGYNQKCY-UHFFFAOYSA-J | |
| tetrapotassium trihydrate dicarbonate | |
| 165.23 (138.21 Anhydrous) | |
| ACS | |
| Hygroscopic |
| Crystalline | |
| 98.5-101.0% | |
| MFCD00149925 | |
| Soluble in water; Practically insoluble in alcohol | |
| O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O | |
| 330.45 | |
| 98.5 to 101.0% | |
| Odorless | |
| Potassium carbonate sesquihydrate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-529-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only