accessibility menu, dialog, popup

UserName

CAS RN 6381-79-9

OOOOOOH2OH2OH2OKKKK

CAS RN 6381-79-9

IUPAC Name: tetrapotassium trihydrate dicarbonate
Synonyms: tetrapotassium trihydrate dicarbonate
Molecular Weight (g/mol): 330.45
Molecular Formula: C2H6K4O9
InChi Key: WWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILES: O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
Grade 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)