missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Reagent grade, Honeywell

Catalog No. p-7072128
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002555 1 kg
6002556 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002555 Supplier Honeywell Chemicals Supplier No. 3478251KG
Only null left

Reagent Grade,≥98%, powder, −325 mesh

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Quantity 1 kg
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
IUPAC Name dipotassium;carbonate
Formula Weight 138.21g/mol
Percent Purity ≥98%
Grade Reagent
Chemical Name or Material Potassium carbonate
Hazard Statement P261-P305 + P351 + P338
RTECSNumber TS7750000