missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Puratronic™, 99.997% (metals basis)

Catalog No. p-7048547
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1083806 5 g
AA1083814 25 g
AA1083822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1083806 Supplier Thermo Scientific Chemicals Supplier No. 01083806
Only null left

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Beilstein 4267587
Merck Index 14,7619
Solubility Information Freely soluble in water. Insoluble in acetone,and alcohol.
IUPAC Name dipotassium;carbonate
Formula Weight 138.21
Percent Purity 99.997%
Grade Puratronic
Sensitivity Hygroscopic
Density 2.29 g/mL
Assay (metals basis)
Chemical Name or Material Potassium carbonate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-529-3
RTECSNumber TS7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only