missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, Anhydrous, Reagents
Potassium Carbonate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$75.20 - $2789.57
Chemical Identifiers
| CAS | 584-08-7 |
|---|---|
| Molecular Formula | CK2O3 |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00011382 |
| InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
| Synonym | Dipotassium Carbonate, Carbonate of Potash |
| IUPAC Name | dipotassium carbonate |
| SMILES | [K+].[K+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-122
|
Labchem Inc
C227950010KG |
10 kg |
Each for $836.80
|
|
|||||
|
01-337-121
|
Labchem Inc
C227950025A5 |
2.5 kg |
Each for $333.43
|
|
|||||
|
01-337-120
|
Labchem Inc
C2279500500A5 |
500 g |
Each for $75.20
|
|
|||||
|
01-337-119
|
Labchem Inc
C227950050F5 |
50 lb. |
Each for $2,789.57
|
|
|||||
Chemical Identifiers
| 584-08-7 | |
| 138.21 | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| dipotassium carbonate |
| CK2O3 | |
| MFCD00011382 | |
| Dipotassium Carbonate, Carbonate of Potash | |
| [K+].[K+].[O-]C([O-])=O |
Specifications
| Colorless/White | |
| 11.6 | |
| 10 kg | |
| MFCD00011382 | |
| BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
| dipotassium carbonate | |
| Reagent | |
| 2.29 |
| 584-08-7 | |
| Solid | |
| CK2O3 | |
| Dipotassium Carbonate, Carbonate of Potash | |
| [K+].[K+].[O-]C([O-])=O | |
| 138.21 | |
| Poly Pail | |
| Potassium Carbonate, Anhydrous |