missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, Anhydrous, Granular, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P1235500GM

 View more versions of this product

Catalog No. 18612516


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

584-08-7
500 g
MFCD00011382
[K+].[K+].[O-]C([O-])=O
138.21
ACS
Poly Bottle
1
CK2O3
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium carbonate
99%
To Pass Test

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
[K+].[K+].[O-]C([O-])=O
CK2O3
MFCD00011382
dipotassium carbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.