missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, anhydrous, 99%

Catalog No. p-7023257
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1662536 500 g
AAA166250E 2500 g
AAA166250C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1662536 Supplier Thermo Scientific Chemicals Supplier No. A1662536
Only null left

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium carbonate has a wide range of industrial and research applications. The major use of potassium carbonate is the manufacture of chemicals, fertilizers, soap and glass. It is used as a drying agent for solvents especially if there are traces of acids present, and as a scavenger of acids produced in organic synthesis. It is used as a buffering agent during the production of wine. It is also used as a fire suppressant in extinguishers. In organic synthesis, it is an extensively used reagent for the alkylation and arylation reactions on carbon, sulfur, nitrogen, and oxygen centres. It is also used in aldol condensation, Michael reaction, acylation, Wittig reaction, Knoevenagel reaction, elimination reaction, epoxide opening, cyclization, metal-catalyzed cross coupling reaction, hydration, and a wide array of reactions.

Solubility
Freely soluble in water. Insoluble in acetone, and alcohol.

Notes
Potassium carbonate is a hygroscopic and deliquescent solid.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
pH 11.5 to 12.5
Quantity 500 g
Linear Formula K2CO3
Beilstein 4267587
Merck Index 14,7619
Solubility Information Freely soluble in water. Insoluble in acetone,and alcohol.
IUPAC Name dipotassium carbonate
Formula Weight 138.21
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.29
Chemical Name or Material Potassium carbonate, anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-529-3
RTECSNumber TS7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only