missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, ACS, 99.0% min

Catalog No. p-4407724
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1260914 25 g
AA1260922 100 g
AA1260936 500 g
AA12609A1 1 kg
AA12609A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA1260914 Supplier Thermo Scientific Chemicals Supplier No. 01260914
Only null left

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color Yellow
Physical Form Granules
Quantity 25 g
Assay Percent Range 99.0% min.
Beilstein 4267587
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name dipotassium carbonate
Formula Weight 138.21
Grade ACS
Sensitivity Hygroscopic
Density 2.29 g/cm3 at 20°C
Chemical Name or Material Potassium carbonate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-529-3
RTECSNumber TS7750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only