Please login to your online account to display your discounted pricing

Potassium Carbonate, 1.5-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$230.27 - $695.25

Chemical Identifiers

Chemical Name or Material Potassium Carbonate
CAS 6381-79-9
Name Note 1.5-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula K2CO3·1.5H2O
Formula Weight 165.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003722

 
avantor performance
301001
Poly Bottle 500g Each for $230.27

02003723

 
avantor performance
301005
Poly Bottle 2.5kg Each for $695.25
Description & Specifications

Specifications

Formula Weight 165.24
Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Potassium Carbonate
CAS 6381-79-9
Molecular Formula K2CO3·1.5H2O
Name Note 1.5-Hydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Quantity 500g