missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Granular, Purified, Approx. 98%, Spectrum™ Chemical

$318.19 - $13194.97

Chemical Identifiers

CAS 2-3-7758
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18612511
View Documents
Spectrum Chemical Mfg Cor
P1230500GM
500 g Amber Glass Bottle
Each for $318.19
Only null left
 
18612512
View Documents
Spectrum Chemical Mfg Cor
P123025KGGL
2.5 kg Amber Glass Bottle
Each for $1,114.31
Only null left
 
18612513
View Documents
Spectrum Chemical Mfg Cor
P123012KG
12 kg Poly Pail
Each for $2,546.72
Only null left
 
18612514
View Documents
Spectrum Chemical Mfg Cor
P123050KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2-3-7758

Specifications

2-3-7758
500 g
Purified
1
∼98%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.