missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Granular, Purified, Approx. 98%, Spectrum™ Chemical

Catalog No. 18612511
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612511 500 g Amber Glass Bottle
18612512 2.5 kg Amber Glass Bottle
18612513 12 kg Poly Pail
18612514 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612511 Supplier Spectrum Chemical Mfg Cor Supplier No. P1230500GM

Chemical Identifiers

CAS 2-3-7758

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity ∼98%
Grade Purified
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.