missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, For IR spectroscopy, Honeywell Fluka™

for IR spectroscopy

$7558.08 - $7558.08

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60046505
View Documents
Honeywell Chemicals
349196X100G
6 x 100 g
Case of 6 Each for $7,558.08
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

2-3-7758
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
[K+].[Br-]
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030

Specifications

1,383°C
2-3-7758
6 x 100 g
CH3(CH2)3CH3
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Spectroscopy
2.73 g/cm3
734°C (literature)
100.0000%
BrK
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
253877
119
Glass Bottle
Potassium bromide, For IR spectroscopy
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1620 days from date of manufacture

EINECSNumber : 231-830-3

Recommended Storage : Room Temp

SDS
Documents

Documents