missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, For IR spectroscopy, Honeywell Fluka™

Catalog No. 60046505
Change view
Click to view available options
Quantity:
6 x 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046505 6 x 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046505 Supplier Honeywell Chemicals Supplier No. 349196X100G
Only null left

for IR spectroscopy

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Quantity 6 x 100 g
Linear Formula CH3(CH2)3CH3
IUPAC Name potassium bromide
Formula Weight 119
Grade Spectroscopy
Packaging Glass Bottle
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, For IR spectroscopy
Show More Show Less
ShelfLife 1620 days from date of manufacture
EINECSNumber 231-830-3
Recommended Storage Room Temp