missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate/Potassium bromide, 0.1N Standardized Solution
Description
Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Colorless |
| Solubility | Miscible with water |
| Quantity | 1 L |
| Chemical Name or Material | Potassium bromate/Potassium bromide |
Safety and Handling
| Hazard Category | H350 |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only