missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromate, ACS Reagent, Reagents
Potassium Bromate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1278500500A5
Specifications
| Potassium Bromate, ACS Reagent | |
| Colorless/White | |
| Solid | |
| BrKO3 | |
| OCATYIAKPYKMPG-UHFFFAOYSA-M | |
| potassium bromate | |
| ACS | |
| 3.27 |
| 434°C | |
| 1-2-7758 | |
| 500 g | |
| Bromic Acid Potassium Salt | |
| [K+].[O-][Br](=O)=O | |
| 167.00 | |
| Poly Bottle |
Chemical Identifiers
| 1-2-7758 | |
| 167.00 | |
| Bromic Acid Potassium Salt | |
| [K+].[O-][Br](=O)=O |
| BrKO3 | |
| OCATYIAKPYKMPG-UHFFFAOYSA-M | |
| potassium bromate |