missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, ACS Reagent, Reagents

Catalog No. p-200021070
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337871 100 g
01337870 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337871 Supplier Labchem Inc Supplier No. C1278500100C2
Only null left

Potassium Bromate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym Bromic Acid Potassium Salt
IUPAC Name potassium bromate
SMILES [K+].[O-][Br](=O)=O

Specifications

Melting Point 434°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.27
Chemical Name or Material Potassium Bromate, ACS Reagent