Learn More
Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals
$201.25 - $201.25
Chemical Identifiers
| Molecular Formula | BrKO3 |
|---|---|
| Molecular Weight (g/mol) | 167.00 |
| MDL Number | MFCD00011359 |
| InChI Key | OCATYIAKPYKMPG-UHFFFAOYSA-M |
| Synonym | potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat |
| PubChem CID | 23673461 |
| ChEBI | CHEBI:38211 |
| IUPAC Name | potassium;bromate |
| SMILES | [O-]Br(=O)=O.[K+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| BrKO3 | |
| MFCD00011359 | |
| potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
| CHEBI:38211 | |
| [O-]Br(=O)=O.[K+] |
| 167.00 | |
| OCATYIAKPYKMPG-UHFFFAOYSA-M | |
| 23673461 | |
| potassium;bromate |
Specifications
| Colorless | |
| BrKO3 | |
| UN3139 | |
| OCATYIAKPYKMPG-UHFFFAOYSA-M | |
| potassium;bromate | |
| 23673461 | |
| 167 | |
| Liquid |
| 7758-01-2,7732-18-5 | |
| MFCD00011359 | |
| potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
| [O-]Br(=O)=O.[K+] | |
| 167.00 | |
| CHEBI:38211 | |
| 1 U | |
| Potassium bromate Acculute Standard Volumetric Solution, Final Concentration 0.1N |
Safety and Handling
GHS H Statement
H271-H350
May cause fire or explosion
strong oxidizer.
May cause cancer.
P201-P202-P281-P308+P313-P501c
H350
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.
EINECSNumber : 231-829-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only