missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bitartrate, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18605467
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605467 12 kg Poly Pail
18605465 500 g Poly Bottle
18605466 2.5 kg Poly Pail
18605468 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605467 Supplier Spectrum Chemical Mfg Cor Supplier No. P180112KG
Only null left

P1801, 868-14-4, C4H5KO6

Spectrum™ Chemical Potassium Bitartrate, USP is used internally as a purgative. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
IUPAC Name potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 99 to 101%
Grade USP
Packaging Poly Pail