missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bisulfate, Reagent Grade, Reagents

Catalog No. 01337873
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337873 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337873 Supplier Labchem Inc Supplier No. C1277500100C2
Only null left

Potassium Bisulfate, Reagent Grade, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym Potassium Hydrogen Sulfate
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Boiling Point 330°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.32
Chemical Name or Material Potassium Bisulfate, Reagent Grade